Anti-LIX1 Rabbit Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A62025-100UG A62025-50UG
77719-512EA 334.95 USD
77719-512 77719-513
Anti-LIX1 Rabbit Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: LIX1
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR