Anti-MEGF10 Rabbit Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A61881-100UG A61881-50UG
77719-243EA 334.95 USD
77719-243 77719-244
Anti-MEGF10 Rabbit Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: MEGF10
Clonality: Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR