Anti-MEGF10 Rabbit Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A61880-50UG A61880-100UG
77719-242EA 228.38 USD
77719-242 77719-241
Anti-MEGF10 Rabbit Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: MEGF10
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR