Anti-THOC3 Rabbit Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A61860-100UG A61860-50UG
77719-202EA 334.95 USD
77719-202 77719-203
Anti-THOC3 Rabbit Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: THOC3
Clonality: Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR