Anti-CEP95 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A61835-50UL A61835-100UL
77719-155EA 228.38 USD
77719-155 77719-154
Anti-CEP95 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200

Type:
Antigen: CEP95
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR