Anti-C3orf20 Rabbit Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A61486-100UG A61486-50UG
77718-489EA 334.95 USD
77718-489 77718-490
Anti-C3orf20 Rabbit Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: C3orf20
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR