Anti-ACBD4 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A61468-100UG A61468-50UG
77718-454EA 334.95 USD
77718-454 77718-455
Anti-ACBD4 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:500-1:2000, IHC:1:20-1:200

Type:
Antigen: ACBD4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR