Anti-CDH26 Rabbit Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A61326-100UG A61326-50UG
77718-190EA 334.95 USD
77718-190 77718-191
Anti-CDH26 Rabbit Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: CDH26
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR