Anti-MYO18B Rabbit Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A61161-50UG A61161-100UG
77717-885EA 228.38 USD
77717-885 77717-884
Anti-MYO18B Rabbit Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: MYO18B
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR