Anti-IRF2BP1 Rabbit Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A61152-50UG A61152-100UG
77717-867EA 228.38 USD
77717-867 77717-866
Anti-IRF2BP1 Rabbit Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: IRF2BP1
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR