Anti-GLB1L2 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A60938-50UG A60938-100UG
77717-460EA 228.38 USD
77717-460 77717-459
Anti-GLB1L2 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: IHC:1:20-1:200

Type:
Antigen: GLB1L2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR