Anti-SLC22A9 Rabbit Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A60935-50UG A60935-100UG
77717-454EA 228.38 USD
77717-454 77717-453
Anti-SLC22A9 Rabbit Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: SLC22A9
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR