Anti-SLC41A1 Rabbit Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A60856-100UG A60856-50UG
77717-308EA 334.95 USD
77717-308 77717-309
Anti-SLC41A1 Rabbit Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: SLC41A1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR