Anti-RIOX2 Rabbit Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A60843-50UG A60843-100UG
77717-285EA 228.38 USD
77717-285 77717-284
Anti-RIOX2 Rabbit Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: RIOX2
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR