Anti-AMOTL2 Rabbit Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A60830-50UL A60830-100UL
77717-259EA 228.38 USD
77717-259 77717-258
Anti-AMOTL2 Rabbit Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: AMOTL2
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR