Anti-COMMD10 Rabbit Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A60824-100UG A60824-50UG
77717-247EA 334.95 USD
77717-247 77717-248
Anti-COMMD10 Rabbit Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: COMMD10
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR