Anti-L Rabbit Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A60701-100UL A60701-50UL
77717-008EA 334.95 USD
77717-008 77717-009
Anti-L Rabbit Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: L
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Vesicular stomatitis Indiana virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR