Anti-MGEA5 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A60371-100UL A60371-50UL
77716-401EA 334.95 USD
77716-401 77716-402
Anti-MGEA5 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:2000, WB:1:200-1:1000, IHC:1:25-1:100

Type:
Antigen: MGEA5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR