Anti-SERPIND1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A60207-100UL
77716-132EA 334.95 USD
77716-132
Anti-SERPIND1 Rabbit Polyclonal Antibody
Antibodies
IHC:1:50-1:100, IF:1:100-1:500

Type:
Antigen: SERPIND1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR