Anti-cyp1a Rabbit Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A48734-100UG A48734-50UG
77715-186EA 334.95 USD
77715-186 77715-187
Anti-cyp1a Rabbit Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: cyp1a
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR