Anti-SIGMAR1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A48409-50UL A48409-100UL
77714-627EA 228.38 USD
77714-627 77714-626
Anti-SIGMAR1 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, WB:1:500-1:2000, IHC:1:100-1:300

Type:
Antigen: SIGMAR1
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR