Anti-CARD9 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A48406-100UL A48406-50UL
77714-621EA 334.95 USD
77714-621 77714-622
Anti-CARD9 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:10000, IHC:1:100-1:300

Type:
Antigen: CARD9
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR