Anti-TAP2 Rabbit Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A48117-50UG A48117-100UG
77714-104EA 228.38 USD
77714-104 77714-103
Anti-TAP2 Rabbit Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: TAP2
Clonality: Polyclonal
Clone:
Conjucation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR