Anti-API5 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A47492-100UL A47492-50UL
77713-048EA 334.95 USD
77713-048 77713-049
Anti-API5 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:5000, WB:1:500-1:2000

Type:
Antigen: API5
Clonality: Polyclonal
Clone:
Conjucation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR