Anti-BCL2L13 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A47253-50UL A47253-100UL
77712-636EA 228.38 USD
77712-636 77712-635
Anti-BCL2L13 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:2000, WB:1:200-1:1000

Type:
Antigen: BCL2L13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR