Anti-SOX18 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A47146-100UL A47146-50UL
77712-452EA 334.95 USD
77712-452 77712-453
Anti-SOX18 Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: SOX18
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR