Anti-PILRB Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A47002-100UL A47002-50UL
77712-203EA 334.95 USD
77712-203 77712-204
Anti-PILRB Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:2000, IHC:1:20-1:100

Type:
Antigen: PILRB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR