Anti-NPTX1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A46980-50UL A46980-100UL
77712-162EA 228.38 USD
77712-162 77712-161
Anti-NPTX1 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, IHC:1:50-1:200

Type:
Antigen: NPTX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR