Anti-CES2 Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A46445-100UL
77711-210EA 334.95 USD
77711-210
Anti-CES2 Rabbit Polyclonal Antibody
Antibodies
WB:1:500-1:3000

Type:
Antigen: CES2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR