Anti-KRT19 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A46429-100UL A46429-50UL
77711-180EA 334.95 USD
77711-180 77711-181
Anti-KRT19 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:5000, WB:1:500-1:2000, IHC:1:100-1:300

Type:
Antigen: KRT19
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR