Anti-GCG Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A46245-100UL A46245-50UL
77710-852EA 334.95 USD
77710-852 77710-853
Anti-GCG Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:2000, IHC:1:10-1:50

Type:
Antigen: GCG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR