Anti-AKT3 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A46219-100UG A46219-50UG
77689-214EA 334.95 USD
77689-214 77710-810
Anti-AKT3 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: IP:1:200-1:2000

Type:
Antigen: AKT3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR