Anti-TP63 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A46201-50UL A46201-100UL
77710-778EA 228.38 USD
77710-778 77710-777
Anti-TP63 Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: TP63
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR