Anti-HDAC1 Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A46136-100UL
77710-656EA 334.95 USD
77710-656
Anti-HDAC1 Rabbit Polyclonal Antibody
Antibodies
WB:1:500-1:3000, IHC:1:50-1:100

Type:
Antigen: HDAC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR