Anti-FOXP3 Rabbit Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A45982-100UG A45982-50UG
77710-391EA 334.95 USD
77710-391 77710-392
Anti-FOXP3 Rabbit Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: FOXP3
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR