Anti-ZNF395 Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A45971-100UL A45971-50UL
77710-373EA 334.95 USD
77710-373 77710-374
Anti-ZNF395 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, IHC:1:50-1:200

Type:
Antigen: ZNF395
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR