Anti-SLC41A2 Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A45970-100UL A45970-50UL
77710-371EA 334.95 USD
77710-371 77710-372
Anti-SLC41A2 Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: SLC41A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR