Anti-mpt64 Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A45886-100UG A45886-50UG
77689-200EA 334.95 USD
77689-200 77710-224
Anti-mpt64 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:500-1:5000

Type:
Antigen: mpt64
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mycobacterium tuberculosis
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR