Anti-COX17 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A45775-100UL
77710-031EA 334.95 USD
77710-031
Anti-COX17 Rabbit Polyclonal Antibody
Antibodies
WB:1:500-1:3000, IHC:1:50-1:100

Type:
Antigen: COX17
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR