Anti-SULT2B1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A45671-50UL A45671-100UL
77709-846EA 228.38 USD
77709-846 77709-845
Anti-SULT2B1 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:2000, IHC:1:10-1:50

Type:
Antigen: SULT2B1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR