Anti-EFNB1/EFNB2/EFNB3 (phospho-Tyr324) Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A45594-100UL
77709-712EA 404.99 USD
77709-712
Anti-EFNB1/EFNB2/EFNB3 (phospho-Tyr324) Rabbit Polyclonal Antibody
Antibodies
WB:1:500-1:1000, IHC:1:50-1:100

Type:
Antigen: EFNB1/EFNB2/EFNB3 (phospho-Tyr324)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR