Anti-AGER Rabbit Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A45582-50UG A45582-100UG
77709-693EA 228.38 USD
77709-693 77709-692
Anti-AGER Rabbit Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: AGER
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR