Anti-SAA1 Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A45529-100UG A45529-50UG
77689-185EA 334.95 USD
77689-185 77709-602
Anti-SAA1 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: IHC:1:20-1:200

Type:
Antigen: SAA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR