Anti-INHBB Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A45488-100UG A45488-50UG
77689-183EA 334.95 USD
77689-183 77709-531
Anti-INHBB Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:500-1:2000, IHC:1:20-1:200, IF:1:50-1:200

Type:
Antigen: INHBB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR