Anti-CFHR2 Rabbit Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A45447-100UG A45447-50UG
77709-458EA 334.95 USD
77709-458 77709-459
Anti-CFHR2 Rabbit Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: CFHR2
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR