Anti-TPST1 Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A45406-50UL A45406-100UL
77709-387EA 228.38 USD
77709-387 77709-386
Anti-TPST1 Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: TPST1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR