Anti-ARRB1 Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A45297-50UL A45297-100UL
77709-192EA 228.38 USD
77709-192 77709-191
Anti-ARRB1 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:5000, IHC:1:50-1:200

Type:
Antigen: ARRB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR