Anti-SLC20A1 Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A45255-50UL A45255-100UL
77709-116EA 228.38 USD
77709-116 77709-115
Anti-SLC20A1 Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: SLC20A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR