Anti-TGFB1I1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A45177-50UG A45177-100UG
77708-973EA 228.38 USD
77708-973 77708-972
Anti-TGFB1I1 Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: TGFB1I1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR