Anti-VTCN1 Rabbit Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A45162-100UG A45162-50UG
77708-944EA 334.95 USD
77708-944 77708-945
Anti-VTCN1 Rabbit Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: VTCN1
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR